Learn More
Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators
Achieve maximum solute recovery with the Thermo Scientific™Deep Vacuum Oil Pump.
Brand:  Thermo Scientific™ VLP200-230
 View more versions of this product
Product Code. 10214832
  • 5085.30 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Achieve maximum solute recovery with the Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators, designed to work with all Thermo Scientific™ SpeedVac™ concentrating and freeze drying systems.
Features:
  • Three-position gas ballast control optimizes pump performance to suit all applications
  • Large, sideview sight glass allows for convenient checking of proper oil levels at a glance
All Rotary Vane Oil Pumps come standard with:
  • Muffler (OME190 Oil Mist Eliminator)
  • Two Oil Filters (MF190)
  • Quick Drain Kit
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VLP200; 230V 50Hz
195L/min. (at 60Hz); 162L/min. (at 50Hz)
0.75 L
12.7 OD
160 mm
260 mm
470 mm
26 kg
230 V
Deep Vacuum Oil Pump
Vacuum Concentrator
1.5 mtorr/1.95 x10³mbar
6 in.
11 in.
19 in.
57 lb.
230 V 50 Hz
50 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products