missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ DNA LoBind™ Plates
GREENER_CHOICE

DNA LoBind Plates maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding. These plates are the ideal solution for sample preparation and long-term storage.

Brand:  Eppendorf™ 0030603303

260.99 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 13427896

  • 272.00 € / Pack of 80
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results.
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yes
Quantitative Protein Analysis
Autoclavable
White, Clear
Polypropylene
96
Non-sterile
Yes
121°C
PCR Clean (Non-sterile)
85.5 x 127.8 x 14.4 mm
Standard
8 x 12
80 Plates (5 Bags x 16 Plates)
Consumables
350 μL
20 to 300 μL
Conical Bottom (V-well)
For PCR techniques
6000 x g
Colorless
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.