missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Deerfit Lux Gloves

Product Code. 12650923
Click to view available options
Size:
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Protect your hands from the cold. Honeywell Safety Products™ are best for light and medium handling in cold conditions.

  • Leather glove-fine black suede deerskin.
  • Grip improved by buffed suede grain.
  • Fully lined with Thinsulate™ for cold protection.
  • Breathable, flexible, ultra lightweight, does not absorb humidity.
  • Elasticized wrist for support and reduced thermal exchange.
  • Black color makes the glove more soil proof.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EC Category PPE 1, EC Certificate Number: Autocertification, EC Attestation: EC Attestation
Classification Dexterity: 4
Cut Style Gunn
Color Black
Exterior Finish Buffed Suede
Material Leather, Thinsulate
Product Type Gloves
Size 9
For Use With (Application) Army-Defense, Energy, Electricity, Homeland Defense, Logistics, Service, Telecom, Transportation and Utility industries
Includes Elastic strip
Liner Material Thinsulate™
Lining Lined
Resistant To Cold
Length (Metric) 25 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'