Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale (EU), Certified
Bench Scales for Basic Industrial Applications
Specifications
Battery Life 100 Hours with Rechargeable Battery
Battery Type AC Adapter (Included);Rechargeable Battery (Included)
Connection Type Standard RS232 and optional USB Device or Ethernet
Construction Stainless steel indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet
Display Type 7-segment Multi-color Backlit LCD
View More Specs
Description
Description
  • Results are clearly shown using the large, bright backlit LCD display.
  • In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
  • This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
  • Indicator features a stainless steel housing and mounting bracket.
  • Features software for Parts Counting, Checkweighing and Totalization.
  • Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
Specifications
Specifications
100 Hours with Rechargeable Battery
Standard RS232 and optional USB Device or Ethernet
7-segment Multi-color Backlit LCD
Weighing, Checkweighing, Counting, Totalization.
AC Adapter (Included);Rechargeable Battery (Included)
Stainless steel indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet
-10°C to +40°C, 85%RH, non-condensing