Learn More
OHAUS™ Defender™ 3000 Stainless Steel
OHAUS™ Defender™ 3000 Stainless Steel is ideal for general weighing and simple counting applications when working in a damp environment. It features a simple, yet rugged stainless steel design, backlit LCD display, and built-in rechargeable battery operation.
800.70 € - 1248.05 €
Specifications
Battery Life 58 Hours with Rechargeable Battery
Connection Type Easy access communication port with RS232 interface (included)
Construction IP65 stainless steel indicator with hinged front cover and eye bolt mounting holes, stainless steel column and wall bracket, 304 grade stainless steel platform with steel tubular frame, IP67 stainless steel load cell
Display Type Backlit LCD
Protection Class IP 65, NEMA 4X, IP65 stainless steel indicator with hinged front cover and eye bolt mounting holes, IP67 stainless steel load cell
View More Specs
Products 3
Product Code Brand Capacity Model Price Quantity & Availability  
Product Code Brand Capacity Model Price Quantity & Availability  
15106062
View Documents
OHAUS™
83997639
150 kg D32XW150VL-EU This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15116062
View Documents
OHAUS™
83997638
60 kg D32XW60VL-EU This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15196052
View Documents
OHAUS™
83997637
60 kg D32XW60VR-EU This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • The NEMA 4X/IP66 stainless steel indicator is built to endure tough industrial elements.
  • The 58-hour rechargeable battery offers continuous operation for multi-day operations.
  • Durable construction with a 304 stainless steel platform, stainless steel tubular frame, column and indicator bracket, and IP67 stainless steel load cell.
  • Easy-to-read display and basic counting mode allow for simple operation for everyday basic industrial applications.
  • Offered with NTEP or OIML/EC Type approvals (depending on region)
Specifications
Specifications
58 Hours with Rechargeable Battery
IP65 stainless steel indicator with hinged front cover and eye bolt mounting holes, stainless steel column and wall bracket, 304 grade stainless steel platform with steel tubular frame, IP67 stainless steel load cell
IP 65, NEMA 4X, IP65 stainless steel indicator with hinged front cover and eye bolt mounting holes, IP67 stainless steel load cell
Gram, Kilogram, Ounce, Pound, Pound:Ounce
Easy access communication port with RS232 interface (included)
Backlit LCD
-10°C to +40°C