Learn More
OHAUS™ Defender™ 5000 - D52 Bench Scale, Models TD52P QV
OHAUS™ Defender™ 5000 Series multifunctional bench scale is ideal for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
1475.00 € - 1585.00 €
Specifications
Battery Life 20 Hours with Rechargeable Battery
Connection Type Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
Construction ABS plastic (TD52P) indicator, 304-grade stainless steel platform with sand gray powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet
Display Type Backlit LCD
Temperature (Metric) Operating -10°C to +40°C
View More Specs
Products 6
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
15908565
View Documents
OHAUS™
30461554
D52P300RQDV3-M
1585.00 €
1 unit
15918455
View Documents
OHAUS™
30424319
D52P60RQDV3
1475.00 €
1 unit
15928455
View Documents
OHAUS™
30424320
D52P150RQDV3
1475.00 €
1 unit
15938455
View Documents
OHAUS™
30424321
D52P300RQDV3
1475.00 €
1 unit
15988555
View Documents
OHAUS™
30461552
D52P60RQDV3-M
1585.00 €
1 unit
15998555
View Documents
OHAUS™
30461553
D52P150RQDV3-M
1585.00 €
1 unit
Description
Description
  • User-friendly model with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
  • Durable construction with a stainless steel platform, sand gray powder-coated steel frame and aluminum load cell.
  • Defender meets NTEP/Measurement Canada or OIML/EC type approvals (depending on region).
  • Features common industrial modes and supports a 50000-item library with 1000 user profiles for easy data management.
  • Includes GMP/GLP with time/date for result traceability.
Specifications
Specifications
20 Hours with Rechargeable Battery
ABS plastic (TD52P) indicator, 304-grade stainless steel platform with sand gray powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet
-10°C to +40°C
38.500 kg
Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
Backlit LCD
773 x 610 x 1203 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
OHAUS™ Defender™ 5000 - D52 Bench Scale, Models TD52P QV