Learn More
OHAUS™ Defender™ 5000 Washdown - D52 Bench Scale, Models TD52XW TV
OHAUS™ Defender™ 5000 Series multifunctional bench scale is ideal for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
2495.00 € - 2600.00 €
Specifications
Battery Life | 20 Hours with Rechargeable Battery |
---|---|
Connection Type | Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock |
Construction | 304-grade stainless steel platform and frame, stainless steel IP68 indicator, IP67 load cell, non-slip adjustable rubber feet |
Display Type | Backlit LCD |
Protection Class | Stainless Seel IP68 Indicator, IP67 Load Cell |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
15918525
|
OHAUS™
30444828 |
D52XW150WTDV8-GB |
2495.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15918585
|
OHAUS™
30461576 |
D52XW150WTDV8-M |
2600.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15928525
|
OHAUS™
30444829 |
D52XW300WTDV8-GB |
2495.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15928585
|
OHAUS™
30461577 |
D52XW300WTDV8-M |
2600.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15938525
|
OHAUS™
30444830 |
D52XW600WTDV8-GB |
2495.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15938585
|
OHAUS™
30461578 |
D52XW600WTDV8-M |
2600.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15958495
|
OHAUS™
30444801 |
D52XW150WTDV8-EU |
2495.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15968495
|
OHAUS™
30444802 |
D52XW300WTDV8-EU |
2495.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15978495
|
OHAUS™
30444803 |
D52XW600WTDV8-EU |
2495.00 €
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- User-friendly model with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
- Durable construction with a stainless steel platform, frame, IP67 load cell and IP68 indicator.
- Defender meets NTEP/Measurement Canada or OIML/EC Type approvals (depending on region).
- Features common industrial modes and supports a 50,000-item library with 1,000 user profiles for easy data management.
- Incudes GMP/GLP with time/date for result traceability.
Specifications
20 Hours with Rechargeable Battery | |
304-grade stainless steel platform and frame, stainless steel IP68 indicator, IP67 load cell, non-slip adjustable rubber feet | |
Stainless Seel IP68 Indicator, IP67 Load Cell | |
963 x 600 x 1234 mm |
Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock | |
Backlit LCD | |
-10°C to +40°C | |
45.300 kg |