missing translation for 'onlineSavingsMsg'
Learn More

Deguelin, MedChemExpress

MedChemExpress Deguelin, a naturally occurring rotenoid, acts as a chemopreventive agent by blocking multiple pathways like PI3K-Akt, IKK-NF-κB, and MAPK-mTOR-survivin-mediated apoptosis. Deguelin binding to Hsp90 leads to a decreased expression of numerous oncogenic proteins, including MEK1/2, Akt, HIF1α, COX-2, and NF-κB.

71.55 € - 464.40 €

Specifications

Chemical Name or Material Deguelin
CAS 522-17-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H22O6
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16485678
Encompass_Preferred
View Documents
Medchem Express
HY-13425
10 mg
132.30 €
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16451847
Encompass_Preferred
View Documents
Medchem Express
HY-13425
50 mg
464.40 €
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16418377
Encompass_Preferred
View Documents
Medchem Express
HY-13425
5 mg
71.55 €
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Deguelin, a naturally occurring rotenoid, acts as a chemopreventive agent by blocking multiple pathways like PI3K-Akt, IKK-NF-κB, and MAPK-mTOR-survivin-mediated apoptosis. Deguelin binding to Hsp90 leads to a decreased expression of numerous oncogenic proteins, including MEK1/2, Akt, HIF1α, COX-2, and NF-κB.
Specifications

Specifications

Deguelin
Light Yellow
C23H22O6
DMSO : 100 mg/mL (253.54 mM; Need ultrasonic)
394.42
98.0%
Solid
522-17-8
Research
(-)-Deguelin (-)-cis-Deguelin
O=C1[C@]2([H])[C@](COC3=CC(OC)=C(OC)C=C32)([H])OC4=C5C=CC(C)(C)OC5=CC=C14
394.42
Research
Cancer-Kinase/protease
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only