Learn More
Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Brand: Thermo Scientific™ D50213
This item is not returnable.
View return policy
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
- Choice of full or half cartridges
Specifications
550 grains | |
Ultrapure | |
Deionizer Cartridge |
Mixed Bed | |
B-Pure™ |
Barnstead Water Purification Cartridge and Filter Systems
File size: 2.3 MB
Product Content Correction
Product Title
Thermo Scientific™ Deionizer Cartridges >
Spot an opportunity for improvement?Share a Content Correction