Learn More
Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Brand: Thermo Scientific™ D0832
147.19 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
- Choice of full or half cartridges
Specifications
Cartridge for ultrapure and organic removal; mixed-bed, activated carbon; full size | |
Mixed Bed, Activated Carbon | |
Bantam™ Deionizer, B-Pure™ |
785 grains/1000 gal. | |
Ultrapure and Organic Removal | |
Deionizer Cartridge |