Learn More
Thermo Scientific™ Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Brand: Thermo Scientific™ D8815
Description
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
- Choice of full or half cartridges
Specifications
High Temp, Organic Removal Cartridge | |
Activated Carbon | |
B-Pure™ |
5000 gal. (as CaCO3) | |
Organic Removal | |
Deionizer Cartridge |