Learn More
Delgocitinib, MedChemExpress
MedChemExpress Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.
Brand:  Medchem Express HY-109053
Encompass_Preferred
 View more versions of this product
Product Code. 16434268
  • 865.35 € / 10mg
Explore more special offers
Description
Description
Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.
Specifications
Specifications
Delgocitinib
Off-White
C16H18N6O
JTE-052
O=C(N(C[C@@H]1C)[C@]21CN(C3=C(C=CN4)C4=NC=N3)CC2)CC#N
310.35
Research
COVID-19-immunoregulation
1263774-59-9
Research
10 mg
DMSO : ≥ 58 mg/mL (186.89 mM)
310.35
99.75%
Solid