Learn More
Chemsplash Delta Coverall (Sterile Irradiated) Product Code.: 30058449
Estimated Shipment: 25-03-2025
to see stock.
Add to basket
Chemsplash Delta Coverall (Sterile Irradiated)
GREENER_CHOICE
Product Code. 30058449
Small
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Product Code. 30058449
Brand: Chemsplash 2766.400.004
Estimated Shipment: 25-03-2025
to see stock.
Add to basket
Add to basket
Delta 67 Coverall - Sterile Irradiated - is made from our soft comfortable 67 gsm microporous laminate fabric, meeting Cat III Type 5B/6B.
Chemsplash Delta 67 Coverall
  • 67GSM Microporous Laminate Fabric
  • Sterile Irradiated, Double Bagged and Detex Sticker
  • Low Linting Knitted Cuffs with Thumb Loops
  • Elasticated Three Piece Hood
  • Two Way Zip
  • Adhesive Zip Flap
  • Elasticated Back and Ankles
  • Reinforced Bound Seam at Crotch Area
  • Non Linting and Anti-Static
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: The Chemsplash Delta 67 Coverall is made from soft, comfortable 67gsm microporous laminate fabric, meeting Cat III Type 5B/6B. It is Sterile Irradiated, double-bagged, and comes with a Detex sticker for identification. The coverall is compliant with EN1149-5:2008 (Antistatic) and EN1073-2 (Nuclear Particle). Chemsplash Delta is ideal for prolonged wear, offering improved breathability while maintaining an effective hazard barrier. This coverall is suitable for a wide range of applications, with an elasticated back, hood, cuffs, ankles, and reinforced seams for added durability. It conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9.
Greener Choice Claims
  • Environmental benefits include:
    • Recycled Content
Specifications
Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit, Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched
Sterility Sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None
Cleanroom Class Class 100-100,000 (ISO 5-8)
Product Title
Chemsplash Delta Coverall (Sterile Irradiated) >