missing translation for 'onlineSavingsMsg'
Learn More

Demeclocycline hydrochloride, MedChemExpress

Product Code. p-200096905
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16497366

Brand: Medchem Express HY17560

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Demeclocycline hydrochloride is a tetracycline antibiotic; is an antibiotic in the treatment of Lyme disease, acne, and bronchitis.

Demeclocycline hydrochloride is a tetracycline antibiotic

Specifications

Chemical Name or Material Demeclocycline hydrochloride
CAS 64-73-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H22Cl2N2O8
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (199.48 mM; Need ultrasonic) ∣H2O : 20 mg/mL (39.90 mM; Need ultrasonic and warming)
SMILES O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])C[C@]3([H])[C@H](O)C4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4Cl)N.Cl
Molecular Weight (g/mol) 501.31
Formula Weight 501.31
Percent Purity 95.09%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.