missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Density Standard (20°C); UKAS ISO/IEC17025 and ISO 17034 certified, nominal density 0.8556 g/mL (20°C), 500 mL, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C to 150°C
Brand: Paragon Scientific DEN20-05-500ML
This item is not returnable.
View return policy
Description
- Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
- Certified in accordance with primary level ASTM D1480 methodology
- Ensures full compliance to ASTM and IP test method protocols
- Supplied in tamper evident packaging with a 2 year shelf life
- Manufactured in the United Kingdom
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- density standard (20
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : white mineral oil (petroleum) (aerosol)
Nominal values quoted. Certified Values will vary from batch to batch.