missing translation for 'onlineSavingsMsg'
Learn More

Density Standards (15 °C), Paragon Scientific™

Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C

79.95 €

Specifications

Quantity 60 mL
View More Specs

Products 15
Product Code Brand Density Price Quantity & Availability  
Product Code Brand Density Price Quantity & Availability  
13400479
View Documents
Paragon Scientific
DEN15-01
0.6654 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13440479
View Documents
Paragon Scientific
DEN15-02
0.7183 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13420479
View Documents
Paragon Scientific
DEN15-04
0.8111 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13430479
View Documents
Paragon Scientific
DEN15-03
0.7807 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13410479
View Documents
Paragon Scientific
DEN15-05
0.8494 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13470479
View Documents
Paragon Scientific
DEN15-06
0.8648 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13450479
View Documents
Paragon Scientific
DEN15-07
0.8811 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13460479
View Documents
Paragon Scientific
DEN15-08
0.9413 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13480479
View Documents
Paragon Scientific
DEN15-09
0.9823 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13490479
View Documents
Paragon Scientific
DEN15-10
1.0248 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13410489
View Documents
Paragon Scientific
DEN15-11
1.0687 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13420489
View Documents
Paragon Scientific
DEN15-12
1.1280 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13430489
View Documents
Paragon Scientific
DEN15-13
1.1962 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13440489
View Documents
Paragon Scientific
DEN15-14
1.2829 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
13460489
View Documents
Paragon Scientific
DEN15-15
1.6300 g/mL (15°C)
79.95 €
60mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols.
  • Supplied in tamper evident packaging (please refer to standard for shelf life).
  • Manufactured in the United Kingdom and available from stock for immediate dispatch
Specifications

Specifications

60 mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DEN15-01

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity Category 2
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications

Nominal values quoted