Learn More
Ansell™ DermaShield™ 73-721 Series Green Neoprene Gloves
Product Code. 11776693
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
Offers the comfort and flexibility of natural rubber gloves, but neoprene composition provides better chemical resistance. Free of latex proteins and accelerators, it minimizes both Type I and Type IV allergies associated with natural rubber.
  • Made from poly-chloroprene
  • For non-sterile controlled environments
  • Free from latex proteins and accelerators
  • Neoprene coating offers chemical resistance
  • Rolled beaded cuff prevents glove from rolling down
  • Bisque fingertip grip
  • Powder-free
TRUSTED_SUSTAINABILITY
Specifications
Back Material Neoprene
Cuff Material Neoprene
Product Type Gloves
Size 8
Style 73-721
Thickness (Metric) 0.175 mm
Glove Cuff Color Green
Glove Cuff Style Beaded/Rolled
Quantity 200 Pairs
Resistant To Chemical resistance
Series 73-721