missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Desiccator with Knob Lid

Desiccator with Knob Lid includes a studded base ring for improved stability.

Brand:  BRAND™ 65031

182.56 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10728051

  • 216.00 € / Each
Estimated Shipment: 21-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Borosilicate Glass 3.3
  • DIN 12491
  • Precisely ground flat flanges
  • Base ring surface studded to improve stability and help prevent cracks
  • Interchangeable lid
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Borosilicate Glass 3.3
Knob Lid
25.2 mm
Desiccator
150 mm
215 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers