missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Borosilicate Glass Desiccator

Studded base ring surface improves stability and helps to prevent cracks. Interchangeable lid included.

Brand:  BRAND™ 65043

358.80 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10589341

  • 448.00 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Borosilicate glass construction
  • Complies with DIN 12491
  • Precisely ground flat flanges
  • Base ring surface studded to improve stability and help prevent cracks
  • Interchangeable lid.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIN 12491
Desiccator
355 mm
320 mm
Borosilicate Glass
Knob Lid
250 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.