missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Desloratadine
Click to view available options
Quantity:
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
CAS | 100643-71-8 |
Melting Point | 150.0°C |
Molecular Formula | C19H19ClN2 |
Quantity | 1 g |
Solubility Information | Solubility aq. soln.: slightly soluble. Other solubilities: soluble in ethanol, propylene glycol, dmso |
InChI Key | JAUOIFJMECXRGI-UHFFFAOYSA-N |
SMILES | ClC1=CC=C2C(CCC3=CC=CN=C3C2=C2CCNCC2)=C1 |
IUPAC Name | 13-chloro-2-(piperidin-4-ylidene)-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene |
Molecular Weight (g/mol) | 310.83 |
Formula Weight | 310.82 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction