Learn More
Brady™ Destructible Tamper-evident Matte Vinyl Labels
Identify product tampering and prevent one-piece label removal with the B-351 tamper-resistant vinyl label material. Easily print almost anywhere with a Brady™ portable label printer.
Brand:  Brady™ 170857
 View more versions of this product
Product Code. 17891829
  • 50.00 € / Pack of 160
Explore more special offers
Description
Description
  • For use in asset tracking, rating plates, product security and tamper identification
  • Designed to fracture to show signs of product tampering and prevent one-piece removal
  • Made with a permanent adhesive that bonds tightly to almost any surface
Specifications
Specifications
Not Autoclavable
M5-20-351
None
Matte
Vinyl
Thermal Transfer
Non-sterile
BMP51, M511
-40°F to +176°F
2 x 1 in.
1 in.
0.5 in.
RoHS Compliant to 2011/65/EU (Pass 2015/863)
Permanent Acrylic
Black on White
Die-cut
Top
Labels
Asset Tracking, Laboratory Labeling, Security Identification
160 Labels/Cartridge
0.0028 in.
2 in.
2.54 cm