Learn More
Brady™ Destructible Tamper Evident Matte Vinyl Labels for M6/M7 Printers
Tamper-resistant vinyl is designed to fracture easily in order to show signs of product tampering and to prevent on-piece label removal.
Brand:  Brady™ 174601
 View more versions of this product
Product Code. 17809154
  • 54.55 € / Pack of 250
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High performance white matte vinyl with permanent adhesive is ideal for applications that require resistance to product tampering
  • Use caution when removing from liner as material is fragile
  • Service temperature of -40°C to 82°C (-40°F to 180°F)
  • Made of B-351 vinyl material
Specifications
Specifications
Not Autoclavable
M7-31-351
None
Matte
Vinyl
Thermal Transfer
Non-sterile
M710, BMP71
250 Labels
-40°F to +176°F
0.0028 in.
1 x 1.5 in.
1.5 in.
25.4 mm
ASTM D 1000, ASTM D 2979, ASTM G 155
Permanent Acrylic
White
Die-cut
Single-side Printable
Labels
General Identification, Laboratory Labeling, Security Identification
No
Rectangle
-40°C to +80°C
0.071 mm
1 in.
38.1 mm