Learn More
Thermo Scientific™ Detergent Removal Spin Columns & Plates
Rapidly eliminate high concentrations of detergents with minimal sample loss with detergent removal resins. Thermo Scientific HiPPR (High Protein and Peptide Recovery) resin is recommended for protein concentrations up to 100 μg/mL, while Thermo Scientific Pierce resin is suitable for concentrations over 100 μg/mL.
165.00 € - 782.00 €
Specifications
Column Type | Proprietary Resin, Affinity |
---|---|
Stationary Phase | Proprietary Ligand |
Purification Target | Detergent Removal |
Product Code | Brand | Quantity | Product Line | Product Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Product Line | Product Type | Price | Quantity & Availability | |||
10116434
|
Thermo Scientific™
87780 |
10 mL | Pierce™ | Detergent Removal Resin |
463.00 €
10mL |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||
10645585
|
Thermo Scientific™
87776 |
25 x 125 μL Columns | Pierce™ | Spin Column |
248.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||
10499764
|
Thermo Scientific™
87777 |
25 x 500 μL Columns | Pierce™ | Spin Column |
309.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||
10792447
|
Thermo Scientific™
87778 |
5 x 2 mL Columns | Pierce™ | Spin Column |
165.00 €
Pack of 5 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||
11736446
|
Thermo Scientific™
87779 |
5 x 4 mL Columns | Pierce™ | Spin Column |
265.00 €
Pack of 5 |
In Stock
Log in to see stock. |
|||
11368261
|
Thermo Scientific™
88304 |
2 x 96-well Plates | Pierce™ | Spin Plate |
782.00 €
Pack of 2 |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
12301883
|
Thermo Scientific™
88305 |
5 mL | HiPPR™ | Spin Column |
306.00 €
1 set |
In Stock
Log in to see stock. |
|||
12391873
|
Thermo Scientific™
88306 |
24 x 0.1 mL Columns | HiPPR™ | Spin Column |
317.00 €
Pack of 24 |
In Stock
Log in to see stock. |
|||
12201233
|
Thermo Scientific™
88307 |
2 x 96-well Plates | HiPPR™ | Spin Plate |
642.00 €
Pack of 2 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
|||
Description
Detergents and surfactants used to prepare protein and peptide samples can interfere with analysis by ELISA, isoelectric focusing, and mass spectrometry (MS). Removing detergents from peptide samples is especially challenging and critical for MS analysis. Even low concentrations of detergents will contaminate instruments and interfere with column binding, elution, and peptide ionization. Thermo Scientific HiPPR and Pierce detergent removal resins are ideal for rapid removal of common detergents from tryptic digests to improve the results of LC-MS/MS and MALDI-MS analysis.
• Optimized—removes >95% of detergent from samples of proteins or peptides• Fast—sample processing takes less than 15 minutes
• Effective—eliminates detergent-interference in downstream applications like ELISA, isoelectric focusing, and mass spectrometry
Pierce Detergent Removal Resin
Pierce Detergent Removal Resin efficiently binds to and removes high concentrations of detergents from 10 μL to 1 mL samples with minimal sample loss. These microcentrifuge columns come prepacked with various volumes of proprietary resin and are best for processing samples with proteins or peptide concentrations greater than 100 μg/mL.
HiPPR Detergent Removal Spin Column Kit
HiPPR Detergent Removal resin has been optimized for sample concentrations of 1 to 100 μg/mL and is ideal for removing commonly used detergents, including SDS, Triton™ X-100, NP-40 and CHAPS at concentrations of 0.5-1%. The resin is available in pre-filled spin columns, 96-well filter spin plates, and as loose resin for customization.
Available formats:
• Resin slurries—bulk resin supplied with empty 0.8-mL spin columns
• Spin columns—0.1 mL micro-centrifuge columns
• 96-well spin plates—pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification
Specifications
Proprietary Resin, Affinity | |
Proprietary Ligand | |
Detergent Removal |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.