missing translation for 'onlineSavingsMsg'
Learn More

Deuterium chloride, 20% w/w in D{2}O, 99.96% (Isotopic), Thermo Scientific Chemicals

Product Code. 11324558
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7698-05-7
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00044642
InChI Key VEXZGXHMUGYJMC-FTGQXOHASA-N
Synonym deuterium chloride, hydrochloric acid-d, deuterium chloride, 20 wt% in deuterium oxide, chlorohydrogen, hydrochloric acid d, chloro 2h, deuterium chloride, 99 atom % d, deuterium chloride, 1 m in deuterium oxide, deuterium chloride, 0.1 m in deuterium oxide, deuterium chloride, 38 wt% in deuterium oxide
PubChem CID 522634
SMILES Cl

Specifications

Physical Form Liquid
Color Colorless to Yellow
Density 1.20 g/mL
Odor Acidic
Packaging Clear Glass Ampule
Refractive Index 1.373
Sensitivity Light and Moisture
Quantity 5 g
Boiling Point 108°C
UN Number UN1789
Solubility Information Fully miscible in water.
Formula Weight 37.47
Percent Purity ≥99.96%
Concentration or Composition (by Analyte or Components) 20% w/w in D2O
Chemical Name or Material Deuterium chloride, Isotopic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.