Learn More
Fisherbrand™ Deuterium Lamps
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
Brand:  Fisherbrand™ 11957-79
 View more versions of this product
Product Code. 15374078
  • 1020.00 € / Each
Explore more special offers
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Produced in state of the art manufacturing facilities approved by lamp manufacturers
  • Full traceability of manufactured products
  • Lamps available for all major instruments
Specifications
Specifications
Deuterium Lamp
400/185 nm
Waters 2487 long life
UV-A, UV-C
0.5 mm
1 Ea.