Learn More
Fisherbrand™ Deuterium Lamps
Designed for compatibility with a variety of instruments. Fisherbrand™ Deuterium Lamps are individually tested, certified, and guaranteed to the same specifications as OEM.
Brand: Fisherbrand™ 11958-17
664.71 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- Produced in state of the art manufacturing facilities approved by lamp manufacturers
- Full traceability of manufactured products
- Lamps available for all major instruments
Specifications
Deuterium Lamp | |
400/185 nm | |
Jasco UV970 975 (B&C) 1570 1575 2070 2075 3075, MD900 MD1500 MD2000 |
UV | |
0.6 mm | |
1 Ea. |