missing translation for 'onlineSavingsMsg'
Learn More
Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™ Product Code.: 10215164

Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™

Product Code. 10215164
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10215164

Brand: Thermo Scientific Acros 426931000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7789-20-0 | H2O | 20.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 1.1050g/mL
Deuterium (D) 99.75% min.
Refractive Index 1.328
Specific Gravity 1.105
Viscosity 1.25 mPa.s (20°C)
Quantity 100 mL
CAS 7789-20-0
Melting Point 3.8°C
Boiling Point 101.4°C
Molecular Formula H2O
Linear Formula D2O
MDL Number MFCD00044636
Merck Index 15, 2942
Synonym deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o
Solubility Information Solubility in water: miscible, exothermic reaction.
InChI Key XLYOFNOQVPJJNP-ZSJDYOACSA-N
SMILES [2H]O[2H]
IUPAC Name [(²H)oxy](²H)
Molecular Weight (g/mol) 20.03
PubChem CID 24602
ChEBI CHEBI:41981
Formula Weight 20.02
Grade NMR
Chemical Name or Material Deuterium oxide, 99.8 atom % D
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467893 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number426931000
Lot NumberA0467741 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number426931000
Lot NumberA0467120 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog Number426931000
Lot NumberA0465731 Certificate TypeCertificate of Analysis Date03/09/2024 Catalog Number426931000
Lot NumberA0465539 Certificate TypeCertificate of Analysis Date02/09/2024 Catalog Number426931000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Deuterium oxide, for NMR, 99.8 atom % D, AcroSeal™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.