Learn More
Dexamethasone 9,11-epoxide, MedChemExpress
MedChemExpress Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.
Brand: Medchem Express HY-N0348
This item is not returnable.
View return policy
Description
Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.Specifications
Dexamethasone 9,11-epoxide | |
White | |
C22H28O5 | |
DMSO : 100 mg/mL (268.49 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) | |
372.45 | |
99.3% | |
Solid |
24916-90-3 | |
Research | |
50 mg | |
C[C@@]12[C@](C(CO)=O)(O)[C@H](C)C[C@@]1([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@]3(O5)[C@]5([H])C2)=O | |
372.45 | |
Research | |
COVID-19-immunoregulation |