missing translation for 'onlineSavingsMsg'
Learn More

Dextran Sulfate Sodium Salt, Colitis Grade (36,000 to 50,000), MP Biomedicals™

Induces severe colitis (inflammatory bowel disease) in laboratory animals

196.00 € - 4040.00 €

Specifications

Color White
pH 5 to 7.11 (1% aqueous solution)
CAS 9011-18-1
Physical Form Powder
Boiling Point 310°C
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11424352
View Documents
MP Biomedicals
0216011010
10 g
196.00 €
10g
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
 
15722262
View Documents
MP Biomedicals
0216011025
25 g
468.00 €
25g
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
11434352
View Documents
MP Biomedicals
0216011050
50 g
861.00 €
50g
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
12871781
View Documents
MP Biomedicals
0216011080
100 g
1540.00 €
100g
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
 
11454352
View Documents
MP Biomedicals
0216011090
500 g
4040.00 €
500g
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
 
Description

Description

Induces severe colitis (inflammatory bowel disease) in laboratory animals

Dextran sulfate sodium salt, or DSS, is a synthetic sulfated branched polysaccharide derivative of dextran that has multiple uses in biomedical and clinical research. One important property of DSS is to trigger colitis in mice and rats when administered in drinking water. DSS binds to medium-chain-length fatty acids in the colon and induces intestinal inflammation. Colitis-grade DSS (MW = 36,000 to 50,000) from MP Bio is one of the most widely used products based on peer reviewed scientific publications.

  • Induces severe colitis (inflammatory bowel disease)
  • Highest sulfur content – 19%
  • Highest chirality - +104° of specific rotation
  • Lowest pH – 6.2 at 1% solution
Specifications

Specifications

White
9011-18-1
310°C
MFCD00081551
Colitis Grade
<10%
5 to 7.11 (1% aqueous solution)
Powder
92°C
Soluble in water
Used for inducing ulcerative colitis in animals and extensively to study colon cancer developing in relation to colonic inflammation
Plastic Container
Videos
Safety and Handling

Safety and Handling

missing translation for 'storageNote' : Store at Room Temperature (15°C to 30°C)

SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.