missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran sulfate sodium salt, approx. M.W. 500,000, Thermo Scientific Chemicals
59.50 € - 385.92 €
Chemical Identifiers
CAS | 9011-18-1 |
---|---|
Molecular Formula | (C6H7O5)m(C6H7O5)n |
Molecular Weight (g/mol) | 40,000 |
MDL Number | MFCD00081551 |
InChI Key | BLFLLBZGZJTVJG-UHFFFAOYSA-N |
Synonym | benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin |
PubChem CID | 2337 |
ChEBI | CHEBI:116735 |
IUPAC Name | Dextran sulfate sodium salt |
SMILES | *-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10577390
|
Thermo Scientific Acros
433240050 |
5 g | Glass bottle |
59.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10814351
|
Thermo Scientific Acros
433240250 |
25 g | Glass bottle |
142.71 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10656820
|
Thermo Scientific Acros
433241000 |
100 g | Glass bottle |
385.92 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9011-18-1 | |
40,000 | |
BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
2337 | |
Dextran sulfate sodium salt |
(C6H7O5)m(C6H7O5)n | |
MFCD00081551 | |
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
CHEBI:116735 | |
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] |
Specifications
9011-18-1 | |
Powder | |
MFCD00081551 | |
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin | |
BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
Dextran sulfate sodium salt | |
2337 | |
Authentic | |
Dextran sulfate sodium salt |
White to Yellow | |
(C6H7O5)m(C6H7O5)n | |
15,2953 | |
colorless to light yellow | |
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*] | |
40,000 | |
CHEBI:116735 | |
10% max. |
Safety and Handling
Product Identifier
- Dextran sulfate, sodium salt
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title