missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran, Thermo Scientific Chemicals
188.94 € - 531.65 €
Chemical Identifiers
CAS | 9004-54-0 |
---|---|
Molecular Formula | (C6H9O5)n(C6H10O5)m |
Molecular Weight (g/mol) | 504.44 |
MDL Number | MFCD00130935 |
InChI Key | FZWBNHMXJMCXLU-UHFFFAOYNA-N |
Synonym | dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal |
PubChem CID | 4125253 |
IUPAC Name | Dextran |
SMILES | O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10637751
|
Thermo Scientific Acros
406271000 |
100 g | Plastic bottle |
188.94 €
100g |
In Stock
Log in to see stock. |
||||
10145363
|
Thermo Scientific Acros
406275000 |
500 g | Plastic bottle |
531.65 €
500g |
Estimated Shipment: 28-11-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9004-54-0 | |
504.44 | |
FZWBNHMXJMCXLU-UHFFFAOYNA-N | |
4125253 | |
O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O |
(C6H9O5)n(C6H10O5)m | |
MFCD00130935 | |
dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal | |
Dextran |
Specifications
White | |
9004-54-0 | |
2.0% max. | |
(C6H9O5)n(C6H10O5)m | |
MFCD00130935 | |
FZWBNHMXJMCXLU-UHFFFAOYNA-N | |
Dextran | |
4125253 | |
Authentic | |
0.22 to 0.27 (1% sol, 37°C) |
4.5 to 7.0 (6% aq. soln.) | |
Powder | |
60000 to 90000 | |
C6H10O5 | |
dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal | |
O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O | |
504.44 | |
Biochemical | |
7.0% max. | |
Dextran, Low fraction, For biochemistry |
Safety and Handling
Product Identifier
- Dextran
RUO – Research Use Only