missing translation for 'onlineSavingsMsg'
Learn More

Dextran (White to Off-white Powder), Fisher BioReagents

Product Code. 10409313
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
Dextran may be used with Polyethylene Glycol under certain conditions in the fractionation of proteins, cells, virus particles, and other macromolecules. Dextran at neutral pH may be autoclaved.
TRUSTED_SUSTAINABILITY

Specifications

Color White
CAS 9004-54-0
Physical Form Solid
Molecular Formula (C6H9O5)n(C6H10O5)m
MDL Number MFCD00130935
Merck Index 15, 2950
Synonym dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal
Quantity 100 g
InChI Key FZWBNHMXJMCXLU-UHFFFAOYNA-N
SMILES O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
IUPAC Name Dextran
Molecular Weight (g/mol) 504.44
PubChem CID 4125253
Identification Pass Test
Loss on Drying 5% max.
Packaging Glass Bottle
Residue after Ignition 1% max.
Chemical Name or Material Dextran
Show More Show Less
Product Identifier
  • Dextran
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.