missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Dextrin Sepharose™ High Performance Media

Provide convenient affinity purification of recombinant proteins with maltose binding protein (MBP) tags

1035.00 € - 3329.00 €

Specifications

Material Dextrin Sepharose
Particle Size 34 μm
Matrix Affinity Resin
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11570684
View Documents
Cytiva
28-9355-97
25 mL
1035.00 €
25mL
In Stock
to see stock.
Add to basket
 
10566305
View Documents
Cytiva
28-9355-98
100 mL
3329.00 €
100mL
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
 
Description

Description

  • Physiological conditions and mild elution preserves activity of target protein
  • Robust, stable medium allows narrow peaks in elution of target protein
  • Compatible with commonly used aqueous buffers and easily regenerated with 0.5M NaOH
  • Easy scale-up

Specifications

Specifications

Dextrin Sepharose
34 μm
Affinity Resin
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • dextrin sepharose high performance

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.