missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrose, Anhydrous, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
Specifications
CAS | 50-99-7 |
Packaging | Poly Bottle |
Molecular Formula | C6H12O6 |
Quantity | 500 g |
Specific Gravity | 1L = 1.54kg |
Synonym | 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form |
InChI Key | GZCGUPFRVQAUEE-SLPGGIOYSA-N |
SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
IUPAC Name | (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
Molecular Weight (g/mol) | 180.156 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction