missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di(2-thienyl)-1,1,2,2-tetramethyldisilane, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 124733-24-0 |
---|---|
Molecular Formula | C12H18S2Si2 |
Molecular Weight (g/mol) | 282.57 |
MDL Number | MFCD09266191 |
InChI Key | FGYSQQKHYAVZEZ-UHFFFAOYSA-N |
Synonym | di 2-thienyl-1,1,2,2-tetramethyldisilane, 1,1,2,2-tetramethyl-1,2-bis thiophen-2-yl disilane, bis 2-thienyl-1,1,2,2-tetramethyldisilane, amtsi088, 1,1,2,2-tetramethyl-1,2-di 2-thienyl disilane, 1,1,2,2-tetramethyl-1,2-di thiophen-2-yl disilane |
PubChem CID | 14382824 |
IUPAC Name | [dimethyl(thiophen-2-yl)silyl]-dimethyl-thiophen-2-ylsilane |
SMILES | C[Si](C)(C1=CC=CS1)[Si](C)(C)C1=CC=CS1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
124733-24-0 | |
282.57 | |
FGYSQQKHYAVZEZ-UHFFFAOYSA-N | |
14382824 | |
C[Si](C)(C1=CC=CS1)[Si](C)(C)C1=CC=CS1 |
C12H18S2Si2 | |
MFCD09266191 | |
di 2-thienyl-1,1,2,2-tetramethyldisilane, 1,1,2,2-tetramethyl-1,2-bis thiophen-2-yl disilane, bis 2-thienyl-1,1,2,2-tetramethyldisilane, amtsi088, 1,1,2,2-tetramethyl-1,2-di 2-thienyl disilane, 1,1,2,2-tetramethyl-1,2-di thiophen-2-yl disilane | |
[dimethyl(thiophen-2-yl)silyl]-dimethyl-thiophen-2-ylsilane |
Specifications
42°C to 44°C | |
C12H18S2Si2 | |
di 2-thienyl-1,1,2,2-tetramethyldisilane, 1,1,2,2-tetramethyl-1,2-bis thiophen-2-yl disilane, bis 2-thienyl-1,1,2,2-tetramethyldisilane, amtsi088, 1,1,2,2-tetramethyl-1,2-di 2-thienyl disilane, 1,1,2,2-tetramethyl-1,2-di thiophen-2-yl disilane | |
C[Si](C)(C1=CC=CS1)[Si](C)(C)C1=CC=CS1 | |
282.57 | |
282.57 | |
Di(2-thienyl)-1,1,2,2-tetramethyldisilane |
124733-24-0 | |
MFCD09266191 | |
FGYSQQKHYAVZEZ-UHFFFAOYSA-N | |
[dimethyl(thiophen-2-yl)silyl]-dimethyl-thiophen-2-ylsilane | |
14382824 | |
97% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title