missing translation for 'onlineSavingsMsg'
Learn More
Di-n-butyl sebacate, 90+%, Thermo Scientific Chemicals Product Code.: 11483757

Di-n-butyl sebacate, 90+%, Thermo Scientific Chemicals

Product Code. 11483757
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11483757

Brand: Thermo Scientific Alfa Aesar A18231.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-43-3
Molecular Formula C18H34O4
Molecular Weight (g/mol) 314.466
MDL Number MFCD00027218
InChI Key PYGXAGIECVVIOZ-UHFFFAOYSA-N
Synonym dibutyl sebacate, butyl sebacate, di-n-butyl sebacate, decanedioic acid, dibutyl ester, polycizer dbs, kodaflex dbs, dibutyl sebacinate, staflex dbs, sebacic acid, dibutyl ester, monoplex dbs
PubChem CID 7986
IUPAC Name dibutyl decanedioate
SMILES CCCCOC(=O)CCCCCCCCC(=O)OCCCC

Specifications

Melting Point -11°C
Density 0.94
Boiling Point 344°C to 345°C
Flash Point 178°C (352°F)
Refractive Index 1.441
Quantity 500 g
Beilstein 1798308
Formula Weight 314.47
Percent Purity 94%
Chemical Name or Material Di-n-butyl sebacate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10239835 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberA18231.36,A18231.22
Lot Number10239744 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog NumberA18231.36,A18231.22
Lot Number10212476 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18231.36,A18231.22
Lot Number10146497 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18231.36,A18231.22
Lot Number10211491 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18231.36,A18231.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Di-n-butyl sebacate, 90+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.