missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylamine, 98+%, Thermo Scientific Chemicals

Product Code. 11469153
Click to view available options
Quantity:
500 mL
2500 mL

Di-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.

Solubility
Miscible with water, ether, ethanol and acetone.

Notes
Incompatible with strong oxidizing agents, carbon dioxide, zinc, iron and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-92-2
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00009429
InChI Key JQVDAXLFBXTEQA-UHFFFAOYSA-N
Synonym dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
PubChem CID 8148
IUPAC Name N-butylbutan-1-amine
SMILES CCCCNCCCC

Specifications

Melting Point °C to 62°C
Density 0.767
Boiling Point 159°C to 160°C
Flash Point 39°C (102°F)
Assay Percent Range ≥98%
Linear Formula [CH3(CH2)3]2NH
Refractive Index 1.4169
Quantity 500 mL
UN Number UN2248
Beilstein 506001
Merck Index 14,3032
Solubility Information Miscible with water,ether,ethanol and acetone.
Formula Weight 129.25
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Di-n-butylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-n-butylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Di-n-butylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'