missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylamine, 99%, Thermo Scientific Chemicals

Product Code. 11935171
Click to view available options
Quantity:
5 mL
50 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Di-n-butylamine, 99%, C8H19N, CAS Number-111-92-2, di-normal-butylamine, n-dibutylamine, n-butyl-1-butanamine, di-n-butyl amine, dibutilamina, dibutylamine, dibutyl amine, di-n-butylamine, dibutyl-amine, 1-butanamine, n-butyl, 2.5L, 98.5% min. (GC), 04, 157, 159.0 deg.C, Undesignated, 0.7600g/mL

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-92-2
MDL Number MFCD00009429
InChI Key JQVDAXLFBXTEQA-UHFFFAOYSA-N
Synonym dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
PubChem CID 8148
IUPAC Name N-butylbutan-1-amine
SMILES CCCCNCCCC

Specifications

Melting Point -62.0°C
Density 0.7600g/mL
Boiling Point 159.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Molecular Formula C8H19N
Linear Formula [CH3(CH2)3]2NH
Refractive Index 1.4160 to 1.4180
Quantity 10 L
Beilstein 04, 157
Fieser 01,209
Merck Index 15, 3037
Specific Gravity 0.76
Solubility Information Solubility in water: 4.05g/L (25°C). Other solubilities: soluble in alcohol, ether, benzene, acetone, miscible with hydrocarbons
Molecular Weight (g/mol) 129.24
Viscosity 0.9 mPa.s (20°C)
Formula Weight 129.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Di-n-butylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-n-butylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'