missing translation for 'onlineSavingsMsg'
Learn More

Di-N-Butylamine, Pure, Fisher Chemical

Product Code. 10345940
Click to view available options
500mL, Plastic Coated Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 111-92-2
Melting Point -62°C
Boiling Point 159°C
Packaging Plastic Coated Glass bottle
Molecular Formula C8H19N
MDL Number 9429
Quantity 500 mL
Vapor Density 4.5
Formula Weight 129.24g/mol
Vapor Pressure 2.3mbar at 20°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-n-butylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'