missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Di-n-butylammonium tetrafluoroborate, 98%
16.00 €
Chemical Identifiers
CAS | 12107-76-5 |
---|---|
Molecular Formula | C8H20BF4N |
Molecular Weight (g/mol) | 217.06 |
MDL Number | MFCD00034899 |
InChI Key | CAZKFGXFAVXKIF-UHFFFAOYSA-O |
Synonym | di-n-butylammonium tetrafluoroborate, dibutylazanium tetrafluoroborate, dibutylamine tetrafluoroborate, dibutylammonium tetrafluoroborate 1-, dibutylammonium tetrafluoroborate, dibutylammonium, tetrafluoroborate, dibutylamine, tetrafluoroborate 1-, 1-butanamine, n-butyl-, tetrafluoroborate 1-, dibutylamine tetrafluoborate, c8h20n.bf4 |
PubChem CID | 25510 |
IUPAC Name | dibutylazanium;tetrafluoroborate |
SMILES | F[B-](F)(F)F.CCCC[NH2+]CCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11489726
|
Alfa Aesar™
B20251.14 |
25 g | N/A | ||||||
Description
Chemical Identifiers
12107-76-5 | |
217.06 | |
CAZKFGXFAVXKIF-UHFFFAOYSA-O | |
25510 | |
F[B-](F)(F)F.CCCC[NH2+]CCCC |
C8H20BF4N | |
MFCD00034899 | |
di-n-butylammonium tetrafluoroborate, dibutylazanium tetrafluoroborate, dibutylamine tetrafluoroborate, dibutylammonium tetrafluoroborate 1-, dibutylammonium tetrafluoroborate, dibutylammonium, tetrafluoroborate, dibutylamine, tetrafluoroborate 1-, 1-butanamine, n-butyl-, tetrafluoroborate 1-, dibutylamine tetrafluoborate, c8h20n.bf4 | |
dibutylazanium;tetrafluoroborate |
Specifications
12107-76-5 | |
(C4H9)2NH2BF4 | |
di-n-butylammonium tetrafluoroborate, dibutylazanium tetrafluoroborate, dibutylamine tetrafluoroborate, dibutylammonium tetrafluoroborate 1-, dibutylammonium tetrafluoroborate, dibutylammonium, tetrafluoroborate, dibutylamine, tetrafluoroborate 1-, 1-butanamine, n-butyl-, tetrafluoroborate 1-, dibutylamine tetrafluoborate, c8h20n.bf4 | |
F[B-](F)(F)F.CCCC[NH2+]CCCC | |
217.06 | |
217.06 | |
Di-n-butylammonium tetrafluoroborate |
C8H20BF4N | |
MFCD00034899 | |
CAZKFGXFAVXKIF-UHFFFAOYSA-O | |
dibutylazanium;tetrafluoroborate | |
25510 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 235-162-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title