missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylgermanium dichloride, 97%, Thermo Scientific Chemicals

Product Code. 11453357
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4593-81-1
Molecular Formula C8H18Cl2Ge
Molecular Weight (g/mol) 257.762
MDL Number MFCD00013590
InChI Key VKMPTHGEGBKPBQ-UHFFFAOYSA-N
Synonym di-n-butylgermanium dichloride, dibutylgermanium dichloride, dichlorodibutylgermane, germane, dibutyldichloro, di-n-butylgermanedichloride, di-n-butyldichlorogermane, dibutyl dichloro germane, wln: g-ge-g4&4
PubChem CID 20723
IUPAC Name dibutyl(dichloro)germane
SMILES CCCC[Ge](CCCC)(Cl)Cl

Specifications

Density 1.208
Boiling Point 241°C to 243°C
Flash Point 107°C (225°F)
Linear Formula [CH3(CH2)3]2GeCl2
Refractive Index 1.473
Quantity 25 g
UN Number UN2922
Sensitivity Moisture sensitive
Formula Weight 257.73
Percent Purity 97%
Chemical Name or Material Di-n-butylgermanium dichloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.