missing translation for 'onlineSavingsMsg'
Learn More
Di-n-butyltin dichloride, 96%, Thermo Scientific Chemicals Product Code.: 11357717

Di-n-butyltin dichloride, 96%, Thermo Scientific Chemicals

Product Code. 11357717
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11357717

Brand: Thermo Scientific Alfa Aesar 014116.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Organotin intermediate

Organotin intermediate.Di-n-butyltin dichloride is used as an organotin intermediate. It serves as heat and light stabilizer for polyvinyl chloride plastics. It is also used in the preparation of methyl 2-(methoxycarbonylmethyl)-5-iodobenzene by reacting with 2,5-diiodobenzoic acid. It acts as an esterification catalyst and a veterinary vermicide and tapeworm remedy. Further, it is used as ion exchange agent, process regulators and processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 683-18-1
Molecular Formula C8H18Cl2Sn
Molecular Weight (g/mol) 303.842
MDL Number MFCD00000518
InChI Key RJGHQTVXGKYATR-UHFFFAOYSA-L
Synonym dibutyltin dichloride, dibutyldichlorotin, di-n-butyltin dichloride, stannane, dibutyldichloro, dibutyltin chloride, dichlorodibutyltin, dibutyl dichloro stannane, dichlorodibutylstannane, dibutyltindichloride, dibutyl tin dichloride
PubChem CID 12688
IUPAC Name dibutyl(dichloro)stannane
SMILES CCCC[Sn](CCCC)(Cl)Cl

Specifications

Melting Point 39°C to 41°C
Color White
Density 1.36g/mL
Boiling Point 135°C (10 mmHg)
Flash Point 168° (334°F)
Odor Acrid
Linear Formula [CH3(CH2)3]2SnCl2
Refractive Index 1.4991
Quantity 50 g
UN Number UN2928
Sensitivity Moisture Sensitive
Solubility Information Insoluble in cold water; Hydrolyzed by hot water; Soluble in organic solvents
Formula Weight 303.83
Percent Purity 96%
Physical Form Crystalline
Chemical Name or Material Di-n-butyltin dichloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Di-n-butyltin dichloride, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.