missing translation for 'onlineSavingsMsg'
Learn More

Di-n-hexyl disulfide, tech. 85%, Thermo Scientific Chemicals

Product Code. 11425147
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10496-15-8
Molecular Formula C12H26S2
Molecular Weight (g/mol) 234.46
MDL Number MFCD00027300
InChI Key GJPDBURPGLWRPW-UHFFFAOYSA-N
Synonym hexyl disulfide, dihexyl disulfide, di-n-hexyl disulfide, disulfide, dihexyl, dihexyl disulphide, 1-hexyldisulfanyl hexane, disulfide,dihexyl, dihexyl persulfide, n-hexyl disulfide, di-n-hexyldisulfide
PubChem CID 82675
IUPAC Name 1-(hexyldisulfanyl)hexane
SMILES CCCCCCSSCCCCCC

Specifications

Density 0.919
Flash Point >110°C (230°F)
Odor Unpleasant
Quantity 5 g
Formula Weight 234.47
Percent Purity 85%
Grade Technical
Chemical Name or Material Di-n-hexyl disulfide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'