missing translation for 'onlineSavingsMsg'
Learn More
Di-n-hexylamine, 98+%, Thermo Scientific Chemicals Product Code.: 11322466

Di-n-hexylamine, 98+%, Thermo Scientific Chemicals

Product Code. 11322466
500 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11322466

Brand: Thermo Scientific Alfa Aesar L06326.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Di-n-hexylamine is used in organic synthesis and involved in the preparation of succinic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-hexylamine is used in organic synthesis and involved in the preparation of succinic acid.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-16-8
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.355
MDL Number MFCD00009521
InChI Key PXSXRABJBXYMFT-UHFFFAOYSA-N
Synonym dihexylamine, di-n-hexylamine, 1-hexanamine, n-hexyl, n,n-dihexylamine, n-hexylhexanamine, unii-k37ada14zv, ccris 4622, k37ada14zv, di-n-heylamine, bis 1-hexyl amine
PubChem CID 8920
IUPAC Name N-hexylhexan-1-amine
SMILES CCCCCCNCCCCCC

Specifications

Melting Point 3°C
Density 0.79
Boiling Point 236°C
Flash Point 104°C (219°F)
Refractive Index 1.4335
Quantity 500 mL
UN Number UN2927
Beilstein 1738519
Solubility Information Miscible with water.
Formula Weight 185.36
Percent Purity ≥98%
Chemical Name or Material Di-n-hexylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0432213 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberL06326.AP,L06326.AE
Lot Number10240436 Certificate TypeCertificate of Analysis Date04/05/2022 Catalog NumberL06326.AP,L06326.AE
Lot Number10045289 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL06326.AP,L06326.AE
Lot Number10231032 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL06326.AP,L06326.AE
Lot Number10162041 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL06326.AP,L06326.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Di-n-hexylamine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.