Learn More
Di-n-octylamine, 96%, Thermo Scientific Chemicals
36.08 € - 76.98 €
Chemical Identifiers
CAS | 1120-48-5 |
---|---|
Molecular Formula | C16H35N |
Molecular Weight (g/mol) | 241.463 |
MDL Number | MFCD00009557 |
InChI Key | LAWOZCWGWDVVSG-UHFFFAOYSA-N |
Synonym | dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine |
PubChem CID | 3094 |
IUPAC Name | N-octyloctan-1-amine |
SMILES | CCCCCCCCNCCCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429156
|
Thermo Scientific Alfa Aesar
A18017.14 |
25 g |
36.08 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439156
|
Thermo Scientific Alfa Aesar
A18017.22 |
100 g |
76.98 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Di-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDi-n-octylamine can be used as intermediate for the preparation of antistatic agent, softener, surfactant and it is also used as metal extraction agent and amine solvent. In addition, it can be used to get octanal.
Solubility
Immiscible in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
Chemical Identifiers
1120-48-5 | |
241.463 | |
LAWOZCWGWDVVSG-UHFFFAOYSA-N | |
3094 | |
CCCCCCCCNCCCCCCCC |
C16H35N | |
MFCD00009557 | |
dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine | |
N-octyloctan-1-amine |
Specifications
1120-48-5 | |
0.799 | |
142°C (287°F) | |
C16H35N | |
1.4432 | |
UN2735 | |
dioctylamine, di-n-octylamine, 1-octanamine, n-octyl, n-n-octyl-n-octylamine, unii-a7hm3062rm, di n-octyl amine, dioctyl-amine, n,n-dioctylamine, di-normal-octylamine | |
LAWOZCWGWDVVSG-UHFFFAOYSA-N | |
N-octyloctan-1-amine | |
3094 | |
98% |
13°C to 15°C | |
297°C to 298°C | |
Amine-like | |
[CH3(CH2)7]2NH | |
MFCD00009557 | |
1748376 | |
Immiscible in water. | |
CCCCCCCCNCCCCCCCC | |
241.463 | |
241.46 | |
Di-n-octylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 214-311-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only