missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyldichlorotin, 95%

Product Code. 11358898
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11358898

Brand: Thermo Scientific Alfa Aesar 041738.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3542-36-7 | C16H36Cl2Sn | 418.07 g/mol

Di-n-octyldichlorotin compounds are used as intermediates in the production of other organotin chemicals. Metatin and Acima. These products are also used as catalysts or industrial intermediates in the production of polyurethanes, silicone resins, synthetic lubricants, and MS polymer sealants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 3542-36-7
Molecular Formula C16H36Cl2Sn
Molecular Weight (g/mol) 418.07
MDL Number MFCD00018091
InChI Key DXPJPPSKHFECLR-UHFFFAOYSA-N
Synonym dichlorodioctyltin, dioctyltin dichloride, di-n-octyltin dichloride, stannane, dichlorodioctyl, dotc, di-n-octyltindichloride, dichloro dioctyl stannane, di-n-octyldichlorotin, stannane, dioctyldichloro, di-n-octyl-zinn dichlorid
PubChem CID 77080
IUPAC Name dichloro(dioctyl)stannane
SMILES Cl.Cl.CCCCCCCC[SnH2]CCCCCCCC

Tekniske data

Melting Point 44°C to 48°C
Boiling Point 175°C (1 mmHg)
Quantity 25 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 416.04
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Di-n-octyldichlorotin

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.