missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyltin oxide, Thermo Scientific Chemicals

Product Code. 11354869
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-08-6
Molecular Formula C16H34OSn
Molecular Weight (g/mol) 361.16
MDL Number MFCD00013839
InChI Key LQRUPWUPINJLMU-UHFFFAOYSA-N
Synonym di-n-octyltin oxide, stannane, dioctyloxo, dioctyltin oxide, dioctyloxostannane, tin, dioctyloxo, dioctyl oxo stannane, stannane, oxodioctyl, di-n-octyl-zinn oxyd, tin, dioctyl-, oxide, dioctyl oxo tin
PubChem CID 93563
IUPAC Name dioctyl(oxo)tin
SMILES CCCCCCCC[Sn](=O)CCCCCCCC

Specifications

Melting Point 245°C to 248°C (decomposition)
Color White
Quantity 25 g
Formula Weight 361.14
Physical Form Powder
Chemical Name or Material Di-n-octyltin oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'