Learn More
Di-n-propyl sulfide, 98+%, Thermo Scientific Chemicals
Description
Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Di-n-propyl sulfide is used in organic synthesis and used as a daily fragrance.
Solubility
Soluble in water 351mg/L @ 25°C.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Specifications
Specifications
Melting Point | -102°C |
Density | 0.838 |
Boiling Point | 141°C to 142°C |
Flash Point | 28°C (82°F) |
Odor | Stench |
Refractive Index | 1.4487 |
Quantity | 500 g |
UN Number | UN1993 |
Beilstein | 1719002 |
Merck Index | 14,7868 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10252842 | Certificate of Analysis | 25/10/2024 | B23155.14,B23155.36,B23155.22 |
10249571 | Certificate of Analysis | 06/02/2024 | B23155.14,B23155.36,B23155.22 |
10246986 | Certificate of Analysis | 24/10/2023 | B23155.14,B23155.36,B23155.22 |
10244785 | Certificate of Analysis | 25/11/2022 | B23155.14,B23155.36,B23155.22 |
10135943 | Certificate of Analysis | 23/11/2021 | B23155.14,B23155.36,B23155.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.