Learn More
Di-p-toluoyl-L-tartaric acid, 98%, Thermo Scientific Chemicals
35.93 € - 353.76 €
Chemical Identifiers
CAS | 32634-66-5 |
---|---|
Molecular Formula | C20H18O8 |
Molecular Weight (g/mol) | 386.36 |
MDL Number | MFCD00064440 |
InChI Key | CMIBUZBMZCBCAT-IYBDPMFKSA-N |
Synonym | 2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid, di-p-toluoyl-l-tartaric acid, l-dtta, --di-p-toluoyl-l-tartaric acid, --di-1,4-o-toluoyl-l-tartaric acid, --o,o'-di-p-toluoyl-l-tartaric acid, l---di-p-toluoyltartaric acid, 2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid, butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r |
PubChem CID | 12297760 |
IUPAC Name | (2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate |
SMILES | CC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11426126
|
Thermo Scientific Alfa Aesar
B22972.06 |
5 g | N/A | ||||||
11436126
|
Thermo Scientific Alfa Aesar
B22972.14 |
25 g |
92.06 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11446126
|
Thermo Scientific Alfa Aesar
B22972.22 |
100 g |
353.76 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
32634-66-5 | |
386.36 | |
CMIBUZBMZCBCAT-IYBDPMFKSA-N | |
12297760 | |
CC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O |
C20H18O8 | |
MFCD00064440 | |
2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid, di-p-toluoyl-l-tartaric acid, l-dtta, --di-p-toluoyl-l-tartaric acid, --di-1,4-o-toluoyl-l-tartaric acid, --o,o'-di-p-toluoyl-l-tartaric acid, l---di-p-toluoyltartaric acid, 2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid, butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r | |
(2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate |
Specifications
32634-66-5 | |
C20H18O8 | |
2022481 | |
CMIBUZBMZCBCAT-IYBDPMFKSA-N | |
(2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate | |
12297760 | |
98% | |
Di-p-toluoyl-L-tartaric acid |
166°C to 171°C | |
MFCD00064440 | |
2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid, di-p-toluoyl-l-tartaric acid, l-dtta, --di-p-toluoyl-l-tartaric acid, --di-1,4-o-toluoyl-l-tartaric acid, --o,o'-di-p-toluoyl-l-tartaric acid, l---di-p-toluoyltartaric acid, 2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid, butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r | |
CC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O | |
386.36 | |
386.36 | |
−139° (c=1 in Ethanol) |
Safety and Handling
missing translation for 'einecsNumber' : 251-131-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.