missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-sec-butoxyaluminoxytriethoxysilane, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Descripción
Di-sec-butoxyaluminoxytriethoxysilane is used as Sol-gel intermediate for aluminum silicates. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
Boiling Point | >200°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 10 g |
UN Number | UN1993 |
Flash Point | 46°C (115°F) |
Solubility Information | Hydrolyzes with water. |
Formula Weight | 352.48 |
Sensitivity | Moisture sensitive |
Density | 1.00 g/mL |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'